Skip to Content
Merck
All Photos(8)

Documents

ZRB2720

Sigma-Aldrich

Anti-GPR37 Antibody, clone 1E14 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

ETBR-LP-1, Endothelin B receptor-like protein 1, G-protein coupled receptor 37, PAELR, Parkin-associated endothelin receptor-like receptor, Prosaposin receptor GPR37

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

recombinant monoclonal

description

clone 1E14
recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 67.11 kDa
observed mol wt ~75 kDa

purified by

using Protein A

species reactivity

rat, human

packaging

antibody small pack of 25

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... GPR37(2861)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E14 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects GPR37. It targets an epitope within 19 amino acids from the first extracellular domain within the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 19 amino acids from the first extracellular domain within the N-terminal half of human GPR37.

Application

Quality Control Testing

Evaluated by Western Blotting in THP-1 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected GPR37 in THP-1 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected GPR37 in lysate from SH-SY5Y cells and 1:1,000 dilution in lysate from Human brain tissue and Rat brain tissue..

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected GPR37 in Human cerebellum tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected GPR37 in SH-SY5Y cells.

Affinity Binding Assay: A representative lot of this antibody bound GPR37 peptide with a KD of 4.7 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Prosaposin receptor GPR37 (UniProt: O15354; also known as Endothelin B receptor-like protein 1, ETBR-LP-1, G-protein coupled receptor 37, Parkin-associated endothelin receptor-like receptor, PAEL-R, PAELR) is encoded by the GPR37 gene (Gene ID: 2861) in human. GPR37 is an orphan G-protein coupled receptor that is exclusively expressed in the nervous system. It is a multi-pass membrane protein with seven transmembrane domains and is synthesized with a signal peptide (aa 1-26) that is subsequently cleaved off to produce the mature protein. The mature GPR37 is exported from the endoplasmic reticulum to the cell surface, where its long extracellular N-terminus is rapidly and constitutively cleaved by metalloproteinases. It serves as a receptor for the neuroprotective and glioprotective factor prosaposin Ligand binding is shown to induce its endocytosis, followed by an ERK phosphorylation cascade. It is reported to mediate the control of oligodendrocyte differentiation. GPR37 expression and mRNA expression are significantly increased in substantia nigra of sporadic Parkinson s disease cases. The level of ecto-GPR37 peptides is also significantly increased in the cerebrospinal fluid of Parkinson s disease subjects, but not in Alzheimer s disease. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Bolinger, AA., et al. (2023). ACS Chem. Neurosci. 14)18); 3318 3334; Morato, X., et al. (2021). Transl. Neurodegener. 10(1); Article 8; Meyer, RC., et al. (2013). Proc. Natl. Acad. Sci. USA. 110(23); 9529-9534).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service