Skip to Content
Merck
All Photos(14)

Key Documents

ZRB2363

Sigma-Aldrich

Anti-ECM1 Antibody, clone 1C6 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Extracellular matrix protein 1, Secretory component p85

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C6, recombinant monoclonal

description

1C6 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 60.67 kDa
observed mol wt ~65 kDa

purified by

using Protein A

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... ECM1(1893)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C6 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Extracellular matrix protein 1 (ECM1). It targets an epitope within 25 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 25 amino acids from the N-terminal region of human Extracellular matrix protein 1 (ECM1).

Application

Quality Control Testing

Evaluated by Western Blotting in Human testis tissue lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ECM1 in Human testis tissue lysate.

Tested Applications

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ECM1 in human epididymis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound ECM1 peptide with a KD of 3.3 x 10-8 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected ECM1 in MCF-7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Extracellular matrix protein 1 (UniProt: Q16610; also known as Secretory component p85) is encoded by the ECM1 gene (Gene ID: 1893) in human. ECM1 is a secreted glycoprotein that is involved in cell proliferation, angiogenesis, and cell differentiation. It is synthesized with a signal peptide (aa 1-19), which is subsequently cleaved off to produce the mature protein that is involved in endochondral bone formation and serves as a negative regulator of bone mineralization. It is shown to interact with a variety of extracellular and structural proteins, contributing to the maintenance of skin integrity and homeostasis. Overexpression of ECM1 has also been reported in several cancers, including pancreatic cancer, breast cancer, gastric cancer, and hepatocellular cancer. It leads to poor prognosis in cancer patients. Its over-expression is also observed in colorectal cancers (CRC) where it is correlated with tumor size, lymph node status and Tumor, Node, Metastasis (TNM) stage of cancer. Its knockdown suppresses CRC cell growth, migration, and invasion. The occurrence and progression of CRC by ECM1 reportedly mediated through the PI3K/AKT/GSK3β/Snail signaling pathway. Mutations in ECM1 gene have been linked to lipoid proteinosis that is characterized by generalized thickening of skin, mucosae. and certain viscera. Four isoforms of ECM1 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Long, S., et al. (2022). Front. Oncol. 12; 889159; Lee, KM., et al. (2014). Breast Can. Res. 16; Article 479).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Reconstitution

300 µg/mL after reconstitution at 25 µL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service