Skip to Content
Merck
All Photos(12)

Key Documents

ZRB2355

Sigma-Aldrich

Anti-TFG Antibody, clone 1F9 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

TRK-fused gene protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1F9, recombinant monoclonal

description

1F9 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 43.45 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

mouse, rat, human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

Gene Information

human ... TFG(10342)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry. This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1F9 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects TFG. It targets an epitope within 23 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 23 amino acids from the C-terminal region of human TFG.

Application

Quality Control Testing

Evaluated by Western Blotting in A549 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected TFG in A549 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected TFG in lysates from HeLa and C6 cells and 1:10,000 dilution in Mouse brain tissue lysate.

Affinity Binding Assay: A representative lot of this antibody bound TFG peptide with a KD of 3.1 x 10-9 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected TFG in Human kidney tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TFG in HeLa cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Protein TFG (UniProt: Q92734; also known as TRK-fused gene protein) is encoded by the TFG gene (Gene ID: 10342) in human. TFG is a ubiquitous protein that is localized in the endoplasmic reticulum (ER) and it plays a role in the normal dynamic function of ER and its associated microtubules. It is required for secretory cargo traffic from ER to the Golgi apparatus. It is involved in the transport of newly synthesized proteins to the endomembrane system via the Coat Protein complex II (COPII) transport vesicles. It is an essential component of type I interferon (IFN) antiviral response. It serves as a TRAF3-interacting protein that positively regulates the RIG-I-Like receptors  (RLR)-dependent type I IFN antiviral response. It is reported to be essential for the formation of the MAVS-TRAF3-TBK1 complex and activation of TBK1 upon RLR activation. TFG is shown to be essential for the phosphorylation of mTOR on serine 2159, a putative TBK1 phosphoacceptor site. Mutations in TFG gene have been linked to several neurodegenerative diseases, including hereditary motor and sensory neuropathy with proximal dominant involvement, the axonal type of Charcot-Marie-Tooth disease, and hereditary spastic paraplegia. Four isoforms of TFG have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Yamamotoya, T., et al. (2022). Sci. Rep. 12; Article 1966; Khan, KA., et al. (2021). PLoS Pathol. 17(1); e1009111; Harlalka, GV., et al. (2016). Hum. Mutat. 37(11); 1157-1161; Witte, K., et al. (2011). Nat. Cell Biol. 13(5); 550-558).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 µL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 µL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service