Skip to Content
Merck
All Photos(9)

Key Documents

ZRB2146

Sigma-Aldrich

Anti-Cyclophilin B Antibody, clone 4D5 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Peptidyl-prolyl cis-trans isom+C4erase B;EC:5.2.1.8;PPIase B;CYP-S1;Rotamase B;S-cyclophilin; SCYLP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4D5, recombinant monoclonal

description

4D5 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 23.74 kDa
observed mol wt ~18 kDa

purified by

using Protein A

species reactivity

human, monkey, rat, mouse

species reactivity (predicted by homology)

porcine, bovine, feline, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CYPB(5479)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies. Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4D5 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Cyclophilin B. It targets an epitope within 17 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the C-terminal region of human Cyclophilin B.

Application

Quality Control Testing

Evaluated by Western Blotting in NIH3T3 cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Cyclophilin B in NIH3T3 cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Cyclophilin B in lysates from COS-7, C6, A431, and U87 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Cyclophilin B in human thyroid tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound recombinant Human Cyclophilin B with a KD of 3.6 x 10-6 in an affinity binding assay.

Enzyme Immunoassay Analysis (ELISA): A representative lot of this antibody detected recombinant Human Cyclophilin B in ELISA application.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Peptidyl-prolyl cis-trans isomerase B (UniProt: P23284; also known as EC:5.2.1.8, PPIase B, CYP-S1, Cyclophilin B, Rotamase B, S-cyclophilin, SCYLP) is encoded by the CYPB (also known as PPIB) gene (Gene ID: 5479) in human. Cyclophilins are a group of peptidyl-prolyl cis/trans isomerases that play crucial role in regulatory mechanisms of cellular physiology and pathology in several inflammatory conditions. Cyclophilin B (CypB) is an endoplasmic reticulum (ER) resident protein that is synthesized with a signal peptide (aa 1-33), which is subsequently cleaved off to produce mature active protein (aa 34-216). It catalyzes the cis-trans isomerization of proline imidic peptide bonds in oligopeptides and therefore assists in protein folding. CypB is secreted by several cell types, and it possesses proinflammatory properties. It is reported to play a key role in ER redox homeostasis. Higher levels of CypB are observed in inflammatory processes, like sepsis and rheumatoid arthritis. It also promotes platelet adhesion to collagen and the neutrophil chemotaxis. It binds to the CD147 membrane receptor to modulate inflammatory pathways. CypB is reported to provide essential survival signals in glioblastoma multiforme cells. Suppression of CypB is shown to reduce the proliferation of glioblastoma cells. Depletion or pharmacological inhibition of CypB causes hyperactivation of the oncogenic RAS-mitogen-activated protein kinase pathway, induction of cellular senescence signals, and death resulting from loss of MYC, mutant p53, Chk1, and JAK/STAT3 signaling. Mutations in CYPB gene are known to cause Osteogenesis imperfecta, a disease characterized by low bone mass, bone fragility and increased susceptibility to fractures even after minimal trauma. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Gegunde, S., et al. (2021). Front. Immunol. 12; 609196; Choi, JW., et al. (2014). Cancer Res. 74(2); 484-496).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service