ZRB2129
Anti-SLC1A3/EAAT1 Antibody, clone 2C3-K ZooMAb® Rabbit Monoclonal


recombinant, expressed in HEK 293 cells
Synonym(s):
Excitatory amino acid transporter 1, Sodium-dependent glutamate/aspartate transporter 1, Solute carrier family 1 member 3
About This Item
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
2C3-K, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 59.57 kDa
observed mol wt ~56 kDa
purified by
using Protein A
species reactivity
mouse, human
species reactivity (predicted by homology)
rat, hamster, porcine, bovine
packaging
antibody small pack of 25
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
western blot: suitable
isotype
IgG
epitope sequence
N-terminal cytoplasmic domain
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
Gene Information
human ... SLC1A3(6507)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in mouse brain tissue lysate.
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected SLC1A3/EAAT1 in mouse brain tissue lysate.
Tested Applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected SLC1A3/EAAT1 in lysate from human brain tissue.
Affinity Binding Assay: A representative lot of this antibody bound SLC1A3/EAAT1 peptide with at least ten thousand-fold (10,000X) higher than with non-specific control peptide.
Immunofluorescence Analysis: A 1:1,000 dilution from a representative lot detected SLC1A3/EAAT1 in human cerebellum tissue sections.
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected SLC1A3/EAAT1 in human cerebellum tissue sections.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service