Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1812

Sigma-Aldrich

Anti-RARγ1 Antibody, clone 1C22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,741.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1C22, recombinant monoclonal

description

1C22 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 50.34 kDa
observed mol wt ~55 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

monkey, monkey, bovine, porcine, horse, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RARG(5916)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1C22 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Retinoic Acid Receptor 1 (RAR 1). It targets an epitope within 16 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 16 amino acids from the N-terminal region of human Retinoic Acid Receptor 1 (RAR 1).

Application

Quality Control Testing

Evaluated by Western Blotting in MCF-7 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected RAR 1 in MCF-7 cell lysate.

Tested applications

Affinity Binding Assay: A representative lot of this antibody bound RAR 1 peptide with a KD of 1.1 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:300 dilution from a representative lot detected RAR 1 in MCF-7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Retinoic acid receptor gamma (UniProt: P13631; also known as RAR-gamma, Nuclear receptor subfamily 1 group B member 3) is encoded by the RARG (also known as NR1B3) gene (Gene ID: 5916) in human. RAR 1 is a member of the nuclear hormone receptor family that serves as a receptor for retinoic acid. Three isoforms of RAR have been described: RAR-a , RAR-β, and RAR- that are expressed in temporal- and tissue-specific patterns beginning early in development and continuing throughout life. RAR- shares about 90% homology with RAR-a and RARβ. It is composed of three domains: a modulating N-terminal domain, a DNA-binding domain (aa 90-155) and a C-terminal ligand-binding domain. It contains two NR C4-type Zinc finger domains (aa 90-110 and 126-150). Four different isoforms of RAR- have been described that are produced by alternative splicing and they differ only in their N-terminal regions. RARs form heterodimers with member of the RXR gene family that also has three different isomers of RXRs (RXR-a, RXR-β, RXR- ) and are expressed in distinct patterns. The binding of all-trans-retinoic acid to RAR in an RAR-RXR heterodimer serves to activate the transcription factor complex, which, when bound to specific retinoic acid response elements (RARE) in target genes can recruit additional protein factors that activate or repress gene expression. The activity of the RARs is further modulated by posttranslational modifications. In human epidermis, RAR- is reported to be the predominant RAR that accounts for about 87% of RAR protein and RAR-a makes up the remainder. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Fisher, GJ., et al. (1998). J. Invest. Dermatol. 110(3); 297-300).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service