Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1811

Sigma-Aldrich

Anti-Tissue Factor Antibody, clone 2L10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

R$1,924.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,924.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

2L10, recombinant monoclonal

description

2L10 Clone

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 33.07 kDa
observed mol wt ~45 kDa

purified by

using Protein A

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal extracellular domain

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... F3(2152)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 2L10 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Tissue Factor. It targets an epitope within 24 amino acids from the N-terminal half, extracellular domain.

Immunogen

KLH-conjugated linear peptide corresponding to 24 amino acids from the N-terminal half, extracellular domain of human Tissue Factor.

Application

Quality Control Testing

Evaluated by Western Blotting in A431 cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected Tissue Factor in A431 cell lysate.

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Tissue Factor in SKOV3 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Tissue Factor in HeLa and SH-SY5Y cells.

Affinity Binding Assay: A representative lot of this antibody bound Tissue Factor peptide with a KD of 1.1 x 10-7 in an affinity binding assay.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Tissue Factor in human kidney tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Tissue factor (UniProt: P13726; also known as TF, Coagulation factor III, Thromboplastin, CD142) is encoded by the F3 gene (Gene ID: 2152) in human. Tissue factor (TF) is an integral membrane protein that serves as a high-affinity receptor and cofactor for factor VII/VIIa. It is expressed on perivascular cells and epithelial cells at organ and body surfaces where it forms a hemostatic barrier. It is also reported to provide hemostatic protection to brain, lung, and heart. In normal healthy individuals, levels of TF are very low. Cell-surface levels of TF do not display any directly correlation with its procoagulant activity indicating that TF exists on the cell surface as dormant (encrypted) and active (de-encrypted) form. This allows for a rapid hemostatic response without a need for transcriptional upregulation of the F3 gene. TF normally exists in a partially encrypted state and its de-encryption on circulating monocytes, platelets or endothelial cells by inflammatory mediators can lead to thrombosis. It initiates blood coagulation by forming a complex with circulating factor VII or VIIa and this complex activates factor IX and X by specific, limited proteolysis. TF expression by nonvascular cells plays an essential role in homeostasis; however, TF expression by vascular cells can induce life-threatening intravascular thrombosis in various diseases, such as sepsis, cancer, and atherosclerosis. Inhibition of TF-Factor VIIa activity is shown to reduce coagulation and mortality in animal models of sepsis. Many cancer cells express large amounts of TF, which contributes to tumor angiogenesis and growth. Cancer cells can release TF-positive microparticles into the circulation that can trigger venous thromboembolism. TF on circulating tumor cells can also coat cells with fibrin that traps them within the microvasculature and promote hematogenous metastasis. This ZooMAbZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Caldwell, JA., et al. (2010). Lab. Invest. 90; 953-962; Kasthuri, RS., et al. (2009). J. Clin. Oncol. 27(29); 4834-4838; Mackman, N. (2004). Arterioscler. Thromb. Vasc. Biol. 24(6);1015-22).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service