Skip to Content
Merck
All Photos(18)

Key Documents

ZRB1659

Sigma-Aldrich

Anti-phospho-HSP27 Ser82 Antibody, clone 1O17 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Heat shock protein beta-1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,741.00


Estimated to ship onApril 09, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1O17, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 22.78 kDa
observed mol wt ~30 kDa

purified by

using Protein A

species reactivity

human

species reactivity (predicted by homology)

bovine, horse, mouse, feline, monkey, canine, rat

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer82)

Gene Information

human ... HSPB1(3315)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1O17 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects HSP27 phosphorylated on serine 82.

Immunogen

KLH-conjugated linear peptide corresponding to 11 amino acids surrounding phosphoserine 82 from the N-terminal half of human Heat shock protein 27 (HSP27).

Application

Quality Control Testing

Evaluated by Western Blotting in lysates from HeLa cells overnight serum starved and treated with Anisomycin.

Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected HSP27 phosphorylated on serine 82 in lysates of HeLa cells overnight serum starved and treated with Anisomycin (25 μg/mL; 30 min.).

Tested applications

Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected phospho-HSP27 Ser82 in lysate from UV-treated HeLa cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected phospho-HSP27 Ser82 in human small intestine and human colon cancer tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound phospho-HSP27-ser82 peptide with a KD of 1. x 10-7 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected phospho-HSP27 Ser82 in HeLa cells treated with Anisomycin.

Peptide Inhibition Assay: Target band detection in lysate from HeLa cells treated with Anisomycin was prevented by preblocking of a representative lot with the immunogen phosphopeptide, but not the corresponding non-phosphopeptide.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Heat shock protein beta-1 (UniProt: P04792; also known as HspB1, 28 kDa heat shock protein, Estrogen-regulated 24 kDa protein, Heat shock 27 kDa protein, HSP 27, Stress-responsive protein 27, SRP27) is encoded by the HSPB1 (also known as HSP27, HSP28) gene (Gene ID: 3315) in human. Heat shock proteins (HSPs) are produced during nonlethal stress conditions and protect the organism from severe and sometimes lethal stress. HSPs can also be induced by non-heat related stress factors. HSP27 is a multidimensional protein that acts as a protein chaperone and an antioxidant and plays a role in the inhibition of apoptosis and actin cytoskeletal remodeling. It is detected in all tissues; however, its highest levels are found in the heart and in tissues composed of striated and smooth muscle. It is shown to be mostly cytoplasmic in interphase cells and is shown to colocalize with mitotic spindles in mitotic cells. It translocates to the nucleus during heat shock and resides in sub-nuclear structures known as nuclear splicing speckles and functions to stabilize DNA and/or the nuclear membrane. HSP-27 contains a conserved -crystalline homology domain at its C-terminal. It is phosphorylated upon exposure to protein kinase C activators and heat shock. HSP27 is present at basal levels in cells and tissues and is organized as large oligomers. After heat shock, exposure to chemical stress, or addition of steroid hormones, its synthesis increases several fold. It is phosphorylated by MAPKAP kinase 2/3 via the activation of the p38 MAPK pathway at multiple serine residues. Following phosphorylation, HSP27 reorganizes itself into smaller oligomers that interact with other proteins. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Alderson TR., et al. (2019). Nat. Commun. 10; Article 1068; Vidyasagar, A., et al. (2012). Fibrogenesis Tissue Repair. 5(1); 7; Landry, J., et al. (1992). J. Biol. Chem. 267(2); 794-803).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service