Skip to Content
Merck
All Photos(16)

Key Documents

ZRB1398

Sigma-Aldrich

Anti-RORγ Antibody, clone 1D13 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

R$1,635.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,635.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1D13, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 58.19 kDa
observed mol wt ~58 kDa

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1D13 is a ZooMAb® mouse recombinant monoclonal antibody that specifically detects Nuclear receptor ROR g. It targets an epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 138 amino acids from the N-terminal half of human Nuclear receptor ROR g.

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected ROR g in Jurkat cell lysate.

Tested applications


Western Blotting Analysis: A 1:10,000 dilution from a representative lot detected ROR g in mouse kidney and HepG2 cell lysates.

Flow Cytometry Analysis: 0.1 μg from a representative lot detected ROR g in one million Human peripheral blood mononuclear cells (PBMC).

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ROR g in human liver tissue sections.

Immunocytochemistry Analysis: A 1:100-1,000 dilution from a representative lot detected ROR g in HeLa and Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound ROR g with a KD of 1.0 x 10-12 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-RORg, clone 1D13 ZooMAb®, Cat. No. ZRB1398, is a recombinant Rabbit monoclonal antibody that specifically targets RORg and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Nuclear receptor RORg (UniProt: P51449; also known as Nuclear receptor RZR-gamma, Nuclear receptor subfamily 1 group F member 3, RAR-related orphan receptor C, Retinoid-related orphan receptor-gamma) is encoded by the RORC (also known as NR1F3, RORG, RZRG) gene (Gene ID: 6097) in human. RORg is a nuclear receptor that binds as a monomer to ROR response elements (RORE) containing a single core motif half-site 5′-AGGTCA-3′ preceded by a short A-T-rich sequence. It is a key regulator of cellular differentiation, immunity, peripheral circadian rhythm as well as lipid, steroid, xenobiotics and glucose metabolism. It is considered to have intrinsic transcriptional activity and also has some natural ligands like oxysterols that act as agonists (25-hydroxycholesterol) or inverse agonists (7-oxygenated sterols), enhancing or repressing the transcriptional activity, respectively. It is reported to recruit distinct combinations of cofactors to target gene regulatory regions to modulate their transcriptional expression, depending on the tissue, time, and promoter contexts. RORg also regulates the circadian expression of clock genes such as CRY1, ARNTL/BMAL1 and NR1D1 in peripheral tissues and in a tissue-selective manner. It negatively regulates adipocyte differentiation through the regulation of early phase gene expression. Two isoforms of ROR g have been described that are produced by alternative promoter usage. Isoform 1 is widely expressed in many tissues, including liver and adipose, and highly expressed in skeletal muscle whereas isoform 2 is primarily expressed in immature thymocytes. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service