Skip to Content
Merck
All Photos(24)

Key Documents

ZRB1369

Sigma-Aldrich

Anti-RAR-alpha Antibody, clone 3L14, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Nuclear receptor subfamily 1 group B member 1, Retinoic acid receptor alpha

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,741.00


Estimated to ship onMay 28, 2025


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3L14, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 50.77 kDa
observed mol wt ~58 kDa

species reactivity

human, canine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... RARA(5914)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3L14 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Retinoic Acid Receptor alpha (RAR-alpha). It targets an epitope within 17 amino acids from the N-terminal half.

Immunogen

KLH-conjugated linear peptide corresponding to 17 amino acids from the N-terminal half of human Retinoic Acid Receptor alpha.

Application

Anti-RAR-alpha, clone 3L14 ZooMAb, Cat. No. ZRB1369, a Rabbit monoclonal antibody that targets RAR-alpha and is tested in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RAR-alpha in A431 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RAR-alpha in HeLa, A431, HUVEC, and NTERA2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100-1,000 dilution from a representative lot detected RAR-alpha in human prostate cancer and human testis tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound Retinoic Acid Receptor alpha peptide with a KD of 9.8 x 10-8 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Retinoic acid receptor alpha (UniProt: P10276; also known as RAR-alpha, Nuclear receptor subfamily 1 group B member 1) is encoded by the RARA (also known as NR1B1) gene (Gene ID: 5914) in human. RAR-alpha is a nuclear receptor of the steroid/thyroid superfamily of nuclear receptor proteins that regulate gene expression by binding to RNA and other elements of DNA response. RAR-alpha forms heterodimer with RXR (via C-terminus) and this association is enhanced by pulsatile shear stress. The RAR-alpha/RXR heterodimer bind to the retinoic acid response elements composed of tandem 5′-AGGTCA-3′ sites known as DR1-DR5. Its nuclear localization depends on ligand binding, phosphorylation, and sumoylation. Phosphorylation on serine 77 is shown to be crucial for its transcriptional activity. Phosphorylation by Akt1 is required for the repressor activity but it has no effect on DNA binding, protein stability, or subcellular localization. In the absence of ligand, the heterodimer associates with a multiprotein complex containing transcription corepressors that induces histone deacetylation, chromatin condensation and transcriptional suppression. Upon ligand binding, the corepressors dissociate from the receptors and associate with the coactivators leading to transcriptional activation. RAR-alpha has two NR C-4 type of zinc-finger domains (aa 88-108 and 124-148). This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service