Skip to Content
Merck
All Photos(26)

Key Documents

ZRB1314

Sigma-Aldrich

Anti-LONP1 Antibody, clone 1I18, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:3.4.21.53, LONHs, LONP, Lon protease homolog mitochondrial, Lon protease-like protein, Mitochondrial ATP-dependent protease Lon, Serine protease 15

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,480.00
4 X 25 μL
R$3,052.00

R$1,480.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,480.00
4 X 25 μL
R$3,052.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.84

R$1,480.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1I18, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 106.49 kDa
observed mol wt ~106 kDa

species reactivity

rat, mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... LONP1(9361)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1I18 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Mitochondrial ATP-dependent protease Lon (LONP).

Immunogen

GST/His-tagged recombinant fragment corresponding to 149 amino acids from the central region of Human LONP1.

Application

Anti-LONP1, clone 1I18 ZooMAb, Cat. No. ZRB1314, is a recombinant rabbit monoclonal antibody that specifically detects mitochondrial Lon protease homolog (LONP1) and has been tested for use in Immunocytochemistry, and Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected LONP1 in A431, HEK293, and L6 cell lysates.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected LONP1 in human small intestine and rat colon tissue sections.

Immunocytochemistry Analysis: A 1:10-100 dilution from a representative lot detected LONP1 in HeLa, A431, HUVEC, NIH 3T3 and MCF-7 cells.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

Lon protease homolog, mitochondrial (UniProt: P36776; also known as EC:3.4.21.53, LONHs, Lon protease-like protein, LONP, Mitochondrial ATP-dependent protease Lon, Serine protease 15) is encoded by the LONP1 (also known as PRSS15) gene (Gene ID: 9361) in human. LONP is a homohexameric, nuclear encoded, mitochondrial matrix protein that acts as an ATP-dependent serine protease. It is expressed ubiquitously and is present at high levels in the liver, brain, heart, skeletal muscle, placenta. It is synthesized with a transit peptide (aa 1-67) that helps in its import into the matrix where it is cleaved off to generate the mature form. It is organized in a ring with a central cavity and the ATP-binding and proteolytic domains (AP-domain) form a hexameric chamber. Its N-terminal domain (aa 124-368) is crucial for the overall structure of the protein, maintaining it in a conformation allowing its proper functioning. It is highly conserved throughout evolution and is shown to be crucial for the maintenance of mitochondrial homeostasis. LONP mediates the selective degradation of mutant and abnormal proteins in mitochondria. It also has a chaperone function in the assembly of inner membrane protein complexes. It also participates in the regulation of mitochondrial gene expression and in the maintenance of the integrity of the mitochondrial DNA. Its DNA and RNA binding is stimulated by substrate and inhibited by ATP binding. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Pinti, M., et al. (2016). Biochim. Biophys. Acta 1857(8); 1300-1306).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service