Skip to Content
Merck
All Photos(12)

Key Documents

ZRB1248

Sigma-Aldrich

Anti-TCF-4 Antibody, clone 1G4 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Estimated to ship onApril 27, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,741.00


Estimated to ship onApril 27, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1G4, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 67.92 kDa
observed mol wt ~85 kDa

purified by

using Protein A

species reactivity

human, rat

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1G4 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects T-cell factor 4 (TCF-4). It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human T-cell factor 4 (TCF-4).

Application

Quality Control Testing

Evaluated by Western Blotting in Rat Liver tissue lysate.

Western Blotting Analysis: A 1:10,000 dilution of this antibody detected TCF-4 in Rat Liver tissue lysate.

Tested applications

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected TCF-4 in HeLa cells and A431 cells.

ELISA Analysis: A r representative lot of this antibody detected TCF-4 peptide in ELISA application..

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-TCF-4, clone 1G4 ZooMAb®, Cat. No. ZRB1248, recombinant Rabbit monoclonal antibody that specifically targets TCF-4 and is tested for use in ELISA, use in ELISA, Immunocytochemistry, and Western Blotting.

Target description

Transcription factor 7-like 2 (UniProt: Q9NQB0; also known as HMG box transcription factor 4, T-cell-specific transcription factor 4, T-cell factor 4, TCF-4, hTCF-4) is encoded by the TCF7L2 (also known as TCF4) gene (Gene ID: 6934) in human. The mammalian T-cell factor family consists of four members (TCF1 4) that contain a highly conserved high-mobility-group (HMG) domain that is responsible for their binding to DNA. TCF-4 is the most prominent member of this family that is highly expressed in small intestine with highest expression at the top of crypts and a gradient expression from crypt to villus. It is also detected in colon epithelium and colon cancer and in mammary gland carcinoma. However, its expression in fetal small intestine is barely detectable. HMG box of TCF-4 is localized to amino acids 350-418. TCF-4 participates in the Wnt signaling pathway and modulates Myc expression by binding to its promoter in a sequence-specific manner. It acts as a repressor in the absence of catenin b1, and as an activator in its presence. It is shown to activate transcription from promoters with several copies of the Tcf motif 5′-CCTTTGATC-3′ in the presence of catenin b1. Transducin-like enhancer of split 1,2,3, and 4 (TLE1-4) are reported to repress transactivation mediated by TCF-4 and catenin b1. TCF-4 undergoes phosphorylation at Thr 201 or Thr 212 by Nemo-like kinase (NLK), which inhibits its DNA-binding and prevents transcriptional activation of target genes of the canonical Wnt/b-catenin signaling pathway. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Costa, AMS., et al. (2013). PLoS One. 8(7); e67694).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service