Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1235

Sigma-Aldrich

Anti-ATG7 Antibody, clone 1E15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,741.00


Estimated to ship onApril 21, 2025


Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1E15, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~77.96 kDa
observed mol wt ~65 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1E15 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects Autophagy related protein 7 (ATG7). It targets an epitope within 13 amino acids from the N-terminal region. It is expected to react with all tree isoforms of ATG7.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the N-terminal region of human Autophagy-related protein 7 (ATG 7).

Application

Quality Control Testing

Evaluated by Western Blotting in HeLa cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ATG7 in HeLa cell lysate.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected ATG7 in NIH3T3 and C6 cell lysates

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected ATG7 in human placenta tissue sections.

Affinity Binding Assay: A representative lot of this antibody bound ATG7 peptide with a KD of 2.4 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ATG7, clone 1E15 ZooMAb®, Cat. No. ZRB1235, is a recombinant Rabbit monoclonal antibody that specifically targets ATG7 and has been tested for use in Affinity Binding Assay, Immunohistochemistry (Paraffin), and Western Blotting.

Target description

Ubiquitin-like modifier-activating enzyme ATG7 (UniProt: O95352; also known as ATG12-activating enzyme E1 ATG7, Autophagy-related protein 7, APG7-like, hAGP7, Ubiquitin-activating enzyme E1-like protein) is encoded by the ATG7 (also known as APG7L) gene (Gene ID: 10533) in human. ATG7 is the E1-like activating enzyme that is involved in the 2 ubiquitin-like systems required for cytoplasm to vacuole transport and autophagy. It is widely expressed, especially in kidney, liver, lymph nodes and bone marrow. Its expression is reported to be upregulated by the transcription factor HSF1. ATG7 interacts with ATG3 and ATG12 and the ATG3-ATG7 complex plays a role in the conjugation of ATG12 to ATG5. The C-terminal region of ATG7 is shown to be essential for the dimerization and interaction with ATG3 and ATG12. Its N-terminal region contains a FAP motif (aa 15-17), which is essential for the formation of ATG7-ATG3 interaction. ATG7 is essential for hematopoietic stem cell maintenance and its absence from the hematopoietic lineage abolishes development of T and B cells. Three isoforms of ATG7 have been described that are produced by alternative splicing. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Tanida, I., et al. (2012). Autophagy. 8(1); 88-97; Mortensen, M., et al. (2011). J. Exp. Med. 208(3); 455-467).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service