Skip to Content
Merck
All Photos(9)

Documents

ZRB1203

Sigma-Aldrich

Anti-Twist1 Antibody, clone 4L15 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Twist-related protein 1;Class A basic helix-loop-helix protein 38;bHLHa38;H-twist

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

4L15, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 20.95 kDa
observed mol wt ~27 kDa

purified by

using Protein A

species reactivity

mouse, human

species reactivity (predicted by homology)

bovine, porcine, feline, horse, monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

N-terminal

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 4L15 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Twist1. It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Twist1.

Application

Quality Control Testing

Evaluated by Western Blotting in HepG2 cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Twist1 in HepG2 cell nuclear extract..

Tested applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Twist1 in nuclear extracts of HeLa cells and BALB 3T3/A31 cells.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Twist1 in HeLa cells.

Affinity Binding Assay: A representative lot of this antibody bound Twist1 peptide with a KD of 7.0 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Twist-related protein 1 (UniProt: Q15672; also known as Class A basic helix-loop-helix protein 38, bHLHa38, H-twist) is encoded by the TWIST1 (also known as BHLHA38, TWIST) gene (Gene ID: 7291) in human. TWIST1 is a basic helix-loop-helix (bHLH) domain (aa 108-159) containing transcription factor with two nuclear localization sequences (aa 37-40 and 73-77). It can form homodimers or heterodimers in order to bind the Nde E-box element and activate or repress its target genes. It is shown to inhibit myogenesis by sequestrating E proteins. TWIST1 is also reported to represses expression of proinflammatory cytokines such as TNF-a and IL-1b. During development, it is shown to be essential for mesoderm specification and differentiation. It promotes EMT in part by directly repressing E-cadherin expression by recruiting the nucleosome remodeling and deacetylase complex for gene repression and by upregulating Bmi1, AKT2, and YB-1. TWIST1 stability is regulated by phosphorylation of serine 68 by MAP kinases (p38, JNK, and ERK1/2). This phosphorylation prevents ubiquitination-mediated degradation of TWIST1. Heterozygous loss-of-function mutations of the human Twist1 gene is shown to cause several diseases including the Saethre-Chotzen syndrome and Robinow-Sorauf syndrome. Twist1-null mouse embryos die with unclosed cranial neural tubes and defective head mesenchyme, somites and limb buds. TWIST1 expression is also observed in many types of aggressive tumors, including breast cancer, hepatocellular carcinoma, prostate cancer, pancreatic cancer, and esophageal squamous cell carcinoma. It can override oncogene-induced cell senescence and apoptosis and can increase cancer cell resistance to chemotherapeutic agents. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Qin, Q., et al. (2012). Cell Res. 22(1); 90-106; Hong, J., et al. (2011). Cancer Res. 71(11); 3980-3990).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

300 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service