Skip to Content
Merck
All Photos(15)

Key Documents

ZRB1177

Sigma-Aldrich

Anti-Iba1/AIF1 Antibody, clone 16B14, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Anti-AIF-1, Anti-IBA1, Anti-IRT-1, Anti-IRT1

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

R$1,924.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

R$1,924.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

16B14, recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 16.70 kDa
observed mol wt ~16 kDa

species reactivity

human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunofluorescence: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable (peptide)

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... AIF1(199)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 16B14 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Allograft inflammatory factor 1 (AIF1). It targets an epitope within 21 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 21 amino acids from the C-terminal region of human Allograft inflammatory factor 1 (AIF1).

Application

Anti-Iba1/AIF1, clone 16B14 ZooMAb, Cat. No. ZRB1177, is a highly specific recombinant rabbit monoclonal antibody that detects Iba1/AIF1 and has been tested for use in Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Iba1/AIF1 in mouse and human brain tissue lysate.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected Iba1/AIF1 in human bone marrow & human cerebral cortex tissue sections.

Immunofluorescence Analysis: A 1:100 dilution from a representative lot detected Iba1/AIF1 in human cerebral cortex tissue sections.

Note: Actual optimal working dilutions must be determined by the end user as specimens and experimental conditions may vary.

Target description

Allograft inflammatory factor 1 (UniProt: P55008; also known as AIF1, Ionized calcium-binding adapter molecule 1, Protein G1) is encoded by the AIF1 (also known as G1, IBA1) gene (Gene ID: 199) in human. AIF1/IBA1 is a highly conserved cytoplasmic, calcium binding, inflammation responsive scaffold protein that is detected in T-lymphocytes and peripheral blood mononuclear cells. It has also been shown to associate with the actin cytoskeleton at membrane ruffles and at sites of phagocytosis. It is not present in normal arteries, but its expression is induced in vascular smooth muscle cells in response to arterial injury where it enhances growth and promote proliferation in vascular smooth muscle cells through deregulation of the cell cycle by shortening the cell cycle and altering the expression of cyclins. AIF1 is also found in activated macrophages where its expression is augmented by cytokines, particularly interferon gamma. It is also considered as a marker for activated macrophages. Serum AIF-1 levels have been correlated with albuminuria and eGFR in type 2 diabetic subjects and serves as a marker for diabetic nephropathy. Three isoforms of AIF1/IBA1 have been described that are produced by alternative splicing. This clone will react with isoforms 1 and 2 that contain this immunogen sequence. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. Fukui, M., et al. (2012). Diabetes Res. Clin. Pract. 98(1); 146-150; Autieri, MV., and Carbone, CM (2001). Arterioscler. Thromb. Vasc. Biol. 21(9);1421-1426.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose. Normal appearance is a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sang Heon Lee et al.
Advanced functional materials, 32(25) (2022-11-17)
The Utah array powers cutting-edge projects for restoration of neurological function, such as BrainGate, but the underlying electrode technology has itself advanced little in the last three decades. Here, advanced dual-side lithographic microfabrication processes is exploited to demonstrate a 1024-channel

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service