Skip to Content
Merck
All Photos(22)

Key Documents

ZRB1129

Sigma-Aldrich

Anti-ZEB1 Antibody, clone 5C20 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

R$1,741.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,741.00
4 X 25 μL
R$3,591.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,741.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

5C20, recombinant monoclonal

description

recombinant, expressed in HEK 293 cells

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 124.07 kDa
observed mol wt ~150 kDa

purified by

using Protein A

species reactivity

mouse, human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

ELISA: suitable
affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb® antibodies represent an entirely new generation of recombinant monoclonal antibodies.Each ZooMAb® antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb® antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 5C20 is a ZooMAb® Rabbit recombinant monoclonal antibody that specifically detects Zinc finger E-box-binding homeobox 1 (ZEB1). It targets an epitope within 22 amino acids from the C-terminal half. It is expected to react with all isoforms.

Immunogen

KLH-conjugated linear peptide corresponding to 22 amino acids from the C-terminal half of human Zinc finger E-box-binding homeobox 1 (ZEB1).

Application

Quality Control Testing

Evaluated by Western Blotting in Jurkat cell lysate.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected ZEB1 in Jurkat cell lysate.

Tested applications

ELISA Analysis: 300 μg/mL from a representative lot detected ZEB1 in ZEB1 in an ELISA.

Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected ZEB1 in HeLa, NIH3T3, HEPG2, A431, and Jurkat cells.

Affinity Binding Assay: A representative lot of this antibody bound ZEB1 with a KD of 2.8 x 10-6 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-ZEB1, clone 5C20 ZooMAb®, Cat. No. ZRB1129, is a recombinant Rabbit monoclonal antibody that detects ZEB1 and is tested for use in Affinity Binding Assay, ELISA, Immunocytochemistry, and Western Blotting.

Target description

Zinc finger E-box-binding homeobox 1 (UniProt: P37275; also known as NIL-2-A zinc finger protein, Negative regulator of IL2, Transcription factor 8, TCF-8) is encoded by the ZEB1 (also known as AREB6, TCF8) gene (Gene ID: 6935) in human. ZEB1 is a transcription factor that is characterized by the presence of two C2H2-type flanking zinc finger clusters, which are responsible for interaction with paired CACCT(G) E-box-like promoter elements on DNA, and a centrally located POU-like homeodomain, not binding DNA. It is a prime element of a network of transcription factors controlling EMT and has been identified as an important molecule in the regulation of DNA damage, cancer cell differentiation, and metastasis. ZEB1 expression is associated with aggressive behavior, metastasis, treatment resistance and poor prognosis in different tumor types, including breast, pancreatic and lung cancer. In breast cancer, highest ZEB1 expression is observed in the aggressive triple negative and basal types. ZEB1 acts as a transcriptional repressor and inhibits interleukin-2 gene expression. It can enhance or repress the promoter activity of the ATP1A1 gene depending on the quantity of cDNA and on the cell type. This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.(Ref.: Drapela, S., et al., (2020). Front. Mol. Biosci. 7; 36).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. The PBS/trehalose components in the ZooMAb® formulation can have the appearance of a semi-solid (bead like gel) after lyophilization. This is a normal phenomenon. Please follow the recommended reconstitution procedure in the data sheet to dissolve the semi-solid, bead-like, gel-appearing material. The resulting antibody solution is completely stable and functional as proven by full functional testing. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

Reconstitute lyophilized antibody pellet with 25μL of ultrapure water or Phosphate Buffered Saline (PBS). Please refer to our reconstitution protocol and the specific application guidance on the suggested starting dilutions and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service