Skip to Content
Merck
All Photos(12)

Documents

ZRB1112

Sigma-Aldrich

Anti-Helicase MOV-10 Antibody, clone 3B8 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Armitage homolog, Moloney leukemia virus 10 protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

3B8, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 113.67 kDa
observed mol wt ~114 kDa

species reactivity

human

species reactivity (predicted by homology)

bovine

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MOV10(4343)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 3B8 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Helicase MOV-10. It targets an epitope within 20 amino acids from the C-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the C-terminal region of human Helicase MOV-10.

Application

Anti-Helicase MOV-10, clone 3B8 ZooMAb, Cat. No. ZRB1112, is a recombinant Rabbit monoclonal antibody that specifically targets Helicase MOV-10 and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry, and Western Blott
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected Helicase MOV-10 in HepG2 cell lysate.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected Helicase MOV-10 in HepG2 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:10 dilution from a representative lot detected Helicase MOV-10 in human pancreas tissue sections.

Affinity Binding Assay:: A representative lot of this antibody bound Helicase MOV-10 peptide with a KD of 2.4 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Helicase MOV-10 (UniProt: EC:3.6.4.13; also known as Armitage homolog, Moloney leukemia virus 10 protein) is encoded by the MOV10 (also known as KIAA1631) gene (Gene ID: 4343) in human. MOV-10 is an evolutionarily conserved member of the UPF1p family of SF-1 ATP-dependent RNA helicases that serves as a putative 5′ to 3′ RNA helicase and contributes to UPF1 mRNA target degradation by translocation along 3′ UTRs. It is shown to inhibit retrovirus replication and overexpression of MOV-10 is shown to severely impair infectivity of HIV-1 and other lentiviruses. It is required for microRNA (miRNA)-mediated gene silencing by the RNA-induced silencing complex (RISC). MOV-10 is also shown to be interferon-inducible RNA helicase and can exhibit antiviral activity independent of its helicase function, against several positive- and negative-strand RNA viruses by enhancing type -I interferon induction. MOV-10 is reported to severely restrict human LINE1 (L1), Alu, and SVA retrotransposons. It associates with the L1 ribonucleoprotein particle, along with other RNA helicases including DDX5, DHX9, DDX17, DDX21, and DDX39A. MOV-10 is co-enriched in cytoplasmic foci along with TUT4. In developing neurons, it is shown to localize both in nucleus and cytoplasm, however, in adults it is found only in the cytoplasmic. It is also required for embryonic viability and for normal central nervous system development and function. It is reported to play two critical roles in early brain development: suppresses retroelements in the nucleus by directly inhibiting cDNA synthesis and regulates cytoskeletal mRNAs to influence neurite outgrowth. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Goodier, JL., et al. (2012). PLoS Genet 8(10); e1002941; Cuevas RA., et al. (2016). J. Immunol. 196(9); 3877-3886).

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Reconstitution

30 μg/mL after reconstitution at 25 μL per vial. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service