Skip to Content
Merck
All Photos(24)

Key Documents

ZRB1068

Sigma-Aldrich

Anti-PRPF19 Antibody, clone 1K10 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

EC:2.3.2.27, Nuclear matrix protein 200, PRP19/PSO4 homolog, Pre-mRNA-processing factor 19, RING-type E3 ubiquitin transferase PRP19, Senescence evasion factor, hPso4

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,480.00
4 X 25 μL
R$3,052.00

R$1,480.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,480.00
4 X 25 μL
R$3,052.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,480.00


Check Cart for Availability

Request a Bulk Order

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1K10, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 55.18 kDa
observed mol wt ~55 kDa

species reactivity

rat, human, mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... PRPF19(27339)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1K10 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects Pre-mRNA-processing factor 19 (PRPF19). It targets an epitope within 20 amino acids from the N-terminal region.

Immunogen

KLH-conjugated linear peptide corresponding to 20 amino acids from the N-terminal region of human Pre-mRNA-processing factor 19 (PRPF19).

Application

Anti-PRPF19, clone 1K10 ZooMAb, Cat. No. ZRB1068, is a recombinant Rabbit monoclonal antibody that specifically targets PRPF19s and is tested for use in Affinity Binding Assay, Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected PRPF19 in HeLa, Jurkat, and L6 cell lysates.

Affinity Binding Assay: A representative lot of this antibody bound PRPF19 peptide with a KD of 2.4 x 10-6 in an affinity binding assay.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected PRPF19 in HeLa, A431, HUVEC, and NIH 3T3 cells.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected PRPF19 in human colon and human testis tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Pre-mRNA-processing factor 19 (UniProt: Q9UMS4; also known as EC:2.3.2.27, Nuclear matrix protein 200, PRP19/PSO4 homolog, hPso4, RING-type E3 ubiquitin transferase PRP19, Senescence evasion factor, PRPF19) is encoded by the PRPF19 (also known as NMP200, PRP19, SNEV) gene (Gene ID: 27339) in human. PRPF19 is a homotetrameric protein that is ubiquitously expressed. Its expression in shown to be elevated in tumor cell lines. PRPF19 is a ubiquitin-protein ligase which is a core component of several complexes mainly involved pre-mRNA splicing and DNA repair. It is required for pre-mRNA splicing as component of the spliceosome. During assembly of the spliceosome it mediates lysine 63-linked polyubiquitination of the U4 spliceosomal protein PRPF3 and ubiquitination of PRPF3 allows its recognition by the U5 component PRPF8 and stabilizes the U4/U5/U6 tri-snRNP spliceosomal complex. PRPF19 distribution varies with the cell cycle stage. It is shown to be nucleoplasmic in interphase cells and is irregularly distributed in anaphase cells. In the prophase stage it is uniformly distributed and in metaphase stage it is found in the extrachromosomal regions. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service