Skip to Content
Merck
All Photos(13)

Key Documents

ZRB1011

Sigma-Aldrich

Anti-CD3e Antibody, clone 1H4, ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

CD3-epsilon, CD3e, T-cell surface antigen T3/Leu-4 epsilon chain, T-cell surface glycoprotein CD3 epsilon chain

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

R$1,635.00


Available to ship onApril 14, 2025Details


Request a Bulk Order

Select a Size

Change View
25 μL
R$1,635.00
4 X 25 μL
R$3,368.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,635.00


Available to ship onApril 14, 2025Details


Request a Bulk Order

biological source

rabbit (recombinant)

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1H4, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 21.39 kDa
observed mol wt ~21 kDa

species reactivity

human

species reactivity (predicted by homology)

monkey

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

flow cytometry: 1:40-1:100
immunocytochemistry: suitable using 1:100
immunofluorescence: 1:40-1:100
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable using 1:1,000

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... CD3E;T3E(916)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 1H4 is a ZooMAb rabbit recombinant monoclonal antibody that specifically detects CD3e. It targets an epitope within 13 amino acids from the C-terminal, cytoplasmic domain.

Immunogen

KLH-conjugated linear peptide corresponding to 13 amino acids from the C-terminal region of human T-cell surface glycoprotein CD3 epsilon chain (CD3e).

Application

Anti-CD3e, clone 1H4 ZooMAb, Cat. No. ZRB1011, is a recombinant Rabbit Monoclonal Antibody that specifically targets of CD3e T-cell surface glycoprotein and has been tested for use in Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting.
Flow Cytometry Analysis: 0.01 μg from a representative lot detected CD3e in Jurkat cell line.

Immunohistochemistry (Paraffin) Analysis: A 1:40 dilution from a representative lot detected CD3e in human tonsil tissue sections.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected CD3e in Jurkat cell line.

Immunofluorescence Analysis: A 1:40 dilution from a representative lot detected CD3e in human tonsil tissue sections.

Target description

T-cell surface glycoprotein CD3 epsilon chain (UniProt P07766; also known as CD3-epsilon, CD3e, T-cell surface antigen T3/Leu-4 epsilon chain) is encoded by the CD3E (also known as T3E) gene (Gene ID 916) in human. The T cell receptor or TCR is a plasma membrane protein complex responsible for recognizing antigens bound to major histocompatibility complex (MHC) on antigen presenting cells. In 95% of T cells, the TCR is composed of a disulfide-linked alpha and beta dimer, while among 5% of T cells the TCR is composed of a gamma and a delta dimer. T cells are activated when their surface TCRs engage with antigen/MHC complex on antigen presenting cells. Each TCR alpha and beta chain contains a variable (V) and a constant (C) extracellular immunoglobulin-like domain, followed by a transmembrane region and a short cytoplasmic tail. The alpha and beta chains have very short cytoplasmic tail and rely on the CD3 (cluster of differentiation 3) co-receptor complex for transducing signaling further intracellularly. The CD3 co-receptor complex is composed of a CD3 chain, a CD3 chain, and two CD3 chains. The CD3 complex, TCR dimer, and a -chain together comprise the functional TCR complex. The CD3 epsilon chain is initially produced with a signal peptide (aa 1-22), the removal of which yields the mature chain (aa. 23-207) with an Ig-like (aa 32-112) domain-containing extracellular region (aa 23-126), a transmembrane segment (aa 127-152), and a cytoplasmic tail (aa 153-207), which contains the immunoreceptor tyrosine-based activation motif or ITAM (a.a. 178-205) that is essential for mediating ZAP-70 interaction and transducing TCR signaling. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals.

Physical form

Purified recombinant rabbit monoclonal antibody IgG, lyophilized in PBS with 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service