Skip to Content
Merck
All Photos(11)

Key Documents

ZMS1037

Sigma-Aldrich

Anti-NG2 Antibody, clone 132.38 ZooMAb® Mouse Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Synonym(s):

Chondroitin sulfate proteoglycan 4, Chondroitin sulfate proteoglycan NG2, HSN tumor-specific antigen

Sign Into View Organizational & Contract Pricing

Select a Size

25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

R$1,924.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 μL
R$1,924.00
4 X 25 μL
R$3,962.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$1,924.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

132.38, recombinant monoclonal
monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt 251.91 kDa
observed mol wt ~300 kDa

species reactivity

rat

species reactivity (predicted by homology)

mouse

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Cspg4(121021)
rat ... Cspg4(81651)

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Learn more about ZooMAb here.

Specificity

Clone 132.38 is a ZooMAb mouse recombinant monoclonal antibody that specifically detects Chondroitin sulfate proteoglycan NG2.

Immunogen

HEK293 cells expressing a truncated integral membrane form of NG2 corresponding to amino acids 1592-2222.

Application

Anti-NG2, clone 132.38 ZooMAb, Cat. No. ZMS1037, is a recombinant Mouse monoclonal antibody that specifically targets NG2 protein and is tested for use in Immunocytochemistry, Immunohistochemistry (Paraffin), and Western Blotting.
Immunocytochemistry Analysis: A 1:1,000 dilution from a representative lot detected NG2 in Oligodendrocytes.

Immunohistochemistry (Paraffin) Analysis: A 1:100 dilution from a representative lot detected NG2 in rat brain tissue sections.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user

Target description

Chondroitin sulfate proteoglycan 4 (UniProt: Q00657; also known as Chondroitin sulfate proteoglycan NG2, HSN tumor-specific antigen) is encoded by the Cspg4 (also known as Ng2) gene (Gene ID: 81651) in rat. Chondroitin sulfate proteoglycan NG2 is a single-pass type I membrane protein that plays a role in cell proliferation and migration and stimulates endothelial cells motility during microvascular morphogenesis. It is present in neural cells and in extraneural tissues, especially in the developing mesenchyme. Its level of expression is highest on immature, proliferating cells and decreases when these cells begin to differentiate. It is found on the surfaces of an unusual class of glial cells within the developing and mature central nervous system that have the properties of oligodendrocyte precursor cells. It is a major chondroitin sulfate proteoglycan that is produced after spinal cord injury and is expressed by macrophages and oligodendrocyte progenitors. It is also reported to inhibit neurite outgrowth and growth cone collapse during axon regeneration. NG2 chondroitin sulfate proteoglycan is synthesized with a signal peptide (aa 1-29), which is subsequently cleaved off in the mature form. This ZooMAb recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. (Ref.: Jones, LL et al. (2002). J. Neurosci. 22(7):2792-2803).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. I am working on multiple sclerosis (MS) using Formalin Fixed Paraffin Embedded human brain tissue. I am very interested in your Anti-NG2 monoclonal antibody (Cat # ZMS1037). Does this antibody work on human brain by immunofluorescent techniques?

    1 answer
    1. This antibody is tested for use in immunohistochemistry (IHC) on paraffin-embedded formalin-fixed in rat brain sections. Unfortunately, this product has not been tested for immunofluorescence or on human samples. While there is some degree of homology, it would be up to the end user to determine suitability. Please see the link below to the UniProt profile to review further information that may be helpful:
      https://www.uniprot.org/uniprotkb/Q00657/entry

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service