Skip to Content
Merck
All Photos(1)

Key Documents

Z768693

Nalgene® media bottles, style 2019

capacity 30 mL

Synonym(s):

bottle nalgene, nalgene containers, sterile bottles

Sign Into View Organizational & Contract Pricing

Select a Size

96 EA
R$2,629.00

R$2,629.00


Check Cart for Availability


Select a Size

Change View
96 EA
R$2,629.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.17

R$2,629.00


Check Cart for Availability

material

square bottle
transparent PETG

Joint

Joints threaded neck (20)

description

Not autoclavable

sterility

sterile; γ-irradiated

feature

graduations

packaging

pack of 96 ea (4 x pack of 24)

manufacturer/tradename

Nalgene® 2019-0030

capacity

30 mL

Looking for similar products? Visit Product Comparison Guide

General description

  • Nalgene® media bottles are made from rigid and sturdy polyethylene terephthalate copolyester, glycol-modified (PETG).
  • Designed for storing buffers, cultures, and culture media for various laboratory applications.
  • Features a leakproof closure for sample protection.
  • Heat-shrink band around closure and neck facilitate tamper-evident seal.
  • Ideal for refrigerated/frozen storage down to -40°C.
Media bottles (square), with white HDPE screw closure. Low gas permeability, non-pyrogenic. Heat-shrink band around closure and neck provides tamper-evident seal.

Features and Benefits

  • Avoids depyrogenation, expensive washing, and autoclaving steps
  • Sterile and non-pyrogenic
  • Obtained in low particulate format
  • Validated by extensive documentation
  • Useful for storing and sampling active pharmaceutical ingredients and bulk intermediates
  • Suitable for preparation and containment of buffers, cultures or extended storage of pH-sensitive liquids such as culture media.

Legal Information

Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service