Skip to Content
Merck
All Photos(1)

Key Documents

Z763837

BeadBug unfilled tubes, with caps and sealing ring

pack of 1000 with caps and sealing ring

Synonym(s):

homogenization microtubes

Sign Into View Organizational & Contract Pricing

Select a Size

1000 EA
R$1,216.00

R$1,216.00


Check Cart for Availability


Select a Size

Change View
1000 EA
R$1,216.00

About This Item

UNSPSC Code:
41106300
NACRES:
NB.22

R$1,216.00


Check Cart for Availability

manufacturer/tradename

Benchmark Scientific D1031-T20

Related Categories

General description

Empty, non-skirted tubes with screw-cap and sealing ring.

For use with BeadBug microtube homogenizer.

Application

BeadBug unfilled tubes, with caps and sealingring, are ideal for use in most bead homogenizers.

Features and Benefits

  • DNAse & RNAse free
  • Non-skirted with a conical bottom

Legal Information

BeadBug is a trademark of Benchmark Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Are the BeadBug™ unfilled tubes (Z763837), along with their caps and sealing ring, RNase free?

    1 answer
    1. The tubes are both DNA and RNase free.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service