Z763438
Orbi-Shaker™ CO2 with remote controller and rubber mat platform
AC/DC input 115 V AC
Synonym(s):
horizontal shaker, lab shaker, reciprocating shaker, remote control laboratory shaker
About This Item
Recommended Products
manufacturer/tradename
Benchmark Scientific BT4001
parameter
30-300 rpm speed
W × L × H
11 in. (28 cm) × 13 in. (33 cm) × 3 in. (7.5 cm)
capacity
5 x 1 L , maximum
platform L × W
13 in. × 13 in.
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
To avoid contamination, unintentional release of gas or deviations from protocol, both speed and time are set via the included remote control module. The remote controller allows the user to set and adjust the shaker settings without opening the chamber door(s).
The instrument is supplied with a 13 x 12 in. non-slip platform.
- Designed for use in C02 incubators & other extreme environments
- Remote controlled, adjust settings from outside the chamber
- Horizontally circular 19 mm orbit for aeration & mixing
- Instantly exchange flask clamps with MAGic Clamp™ accessories
Legal Information
accessory
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service