Z757357
Sartorius Proline® Plus mechanical pippettor
single channel, volume 2-20 μL
Select a Size
R$2,661.00
Select a Size
About This Item
R$2,661.00
Recommended Products
feature
electronic: no
single-channel
packaging
pack of 1 ea
manufacturer/tradename
Sartorius 728030
volume
2-20 μL
Related Categories
General description
- Reduced pipetting forces prevent RSI and improve the results
- Accurate and precise
- Contamination free pipetting with Safe-Cone Filters
- Robust design
- Easiest pipette to maintain
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)