Z744249
Wilmad® 50 ml Suprasil® Small Volume Liquid Nitrogen Dewar
for Bruker®, O.D. 10 mm, pack of 1 ea
Synonym(s):
EPR cryo dewar, EPR dewar
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.15
Recommended Products
packaging
pack of 1 ea
manufacturer/tradename
Wilmad WG-816-B-Q
O.D.
10 mm
General description
Dewar Flasks provide a convenient alternative to Variable Temperature systems for studies at 77 K, and are silvered around the reservoir.
Legal Information
Bruker is a registered trademark of Bruker-Physik AG
Suprasil is a registered trademark of HERAEUS QUARZGLAS GMBH & CO.
Wilmad is a registered trademark of SP Industries
Choose from one of the most recent versions:
Certificates of Analysis (COA)