Z743515
Laser WetGrip labels
white, L × W 0.94 in. × 0.50 in., roll of 380
Synonym(s):
moisture resistant labels
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
packaging
roll of 380
manufacturer/tradename
Diversified Biotech GRLA-2380
L × W
0.94 in. × 0.50 in.
color
white
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Labels for Moist, Frosty or Frozen Surfaces
"Grips" to wet, frost-covered and frozen surfaces
Temperature resistant to -80°C
Compatible with centrifuge rotors
Resistant to chemicals and solvents
Adheres to plastics, glass, metals and even paperboard boxes
Supplied in white laser sheets
"Grips" to wet, frost-covered and frozen surfaces
Temperature resistant to -80°C
Compatible with centrifuge rotors
Resistant to chemicals and solvents
Adheres to plastics, glass, metals and even paperboard boxes
Supplied in white laser sheets
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service