Skip to Content
Merck
All Photos(1)

Key Documents

Z742231

Labnet Fastpette Pro Pipette Controller

AC/DC input 100 - 240 V AC, universal plug set (US, Europlug, UK and Australian Plugs)

Synonym(s):

Fastpette Pro Controller, Labnet Pipette Controller, Pro Pipette Controller

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$3,667.00

R$3,667.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$3,667.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

R$3,667.00


Check Cart for Availability

feature

CE compliant
Fits glass or plastic, 1-100 mL

manufacturer/tradename

Labnet P2002

volume

0.5-100 mL

battery power

3 AAA, 8 hr life (NiMH)

Related Categories

General description

The new FASTPETTE PRO- Lightweight and ergonomically shaped motorized pipette controller designed for work with glass or plastic serological pipettes from 0.5 to 100 ml range. Conveniently positioned switches allow choosing different operation modes and speed depending on the volume of the serological pipette and liquid viscosity. Additionally aspirating and dispensing speed is controlled through two comfortable finger triggers.

Components

  • Charger and stand
  • Autoclavable filter 0.2 um
  • Batteries
  • Universal power supply

Other Notes

When fully charged, the NiMH batteries enable up to eight hours of continuous use. The large LCD display the serological high viscosity faspette indicates the battery status along with the pipetting mode and speed selected. In order to protect the device against overfilling the FASTPETTE PRO is equipped with a 0.2 um PTFE filter. The filter, the pipette holder and the nosepiece are autoclavable. The FASTPETTE PRO is also supplied with a universal battery charger, an additional hydrophobic 0.2 μm filter and a two-position charging stand.

Legal Information

Corning is a registered trademark of Corning, Inc.
FastPette is a trademark of Labnet Intl., Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service