Z740085
BRAND® pipette tips
volume 0.5-5 mL, bulk
Synonym(s):
BRAND tips, micropipette tips, pipette tips, tips
Select a Size
R$480.00
Available to ship onApril 16, 2025Details
Select a Size
About This Item
R$480.00
Available to ship onApril 16, 2025Details
Recommended Products
material
colorless
colorless polypropylene tip
polypropylene
sterility
non-sterile; autoclaved
feature
bulk
packaging
pack of 200 ea (1 bag of 200)
manufacturer/tradename
BRAND 702595
maximum volume
5 mL
volume
0.5-5 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
Features and Benefits
- CERTIFIED LIFE SCIENCE QUALITY: Virgin raw materials, Manufactured under controlled room conditions, Intensive in-process controls, Visual inspections, Batch management, Final product inspection, Proven functionality
- Perfect fit for the Transferpette® micropipette and many other pipettes.
- The tip system for all requirements.
- BRAND pipette tips and filter tips as well as the corresponding TipBoxes are autoclavable at 121 °C (2 bar) acc. DIN EN 285.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service