Skip to Content
Merck
All Photos(1)

Documents

Z721581

Sigma-Aldrich

EZ-Pierce films

Excel Scientific EZP-100, non-sterile

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

material

polyethylene film

sterility

non-sterile

feature

piercable

packaging

pack of 100 ea

manufacturer/tradename

Excel Scientific EZP-100

L

146.1 mm

W × L

8.26 cm × 14.61 cm

W

82.6 mm

Looking for similar products? Visit Product Comparison Guide

General description

  • Easily pierceable.
  • 70 μm thick, polyethylene with acrylic adhesive.
  • EZ-Pierce films allow direct sample recovery with single or multichannel pipettors and robotic probes.
  • Dimensions are 82.6 mm by 142.9 mm for 96 well plates.

Legal Information

EZ-Pierce is a trademark of Excel Scientific, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Novel sample prep technique coupled with the unique selectivity of the Ascentis Express F5 column enables a fast and simplified bioanalytical method for associated vitamin D metabolites.

Using HybridSPE-Phospholipid, Ascentis Express, and Other LC-MS Workflow Components

Protocols

The conversion of clinical methods to LC/MS/MS offers advantages; however, is accompanied by a few limitations, notably interference effects from the endogenous sample matrix.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service