Skip to Content
Merck
All Photos(1)

Key Documents

Z721476

SealPlate® ColorTab films

assorted colors

Sign Into View Organizational & Contract Pricing

Select a Size

50 EA
R$317.00

R$317.00


Check Cart for Availability


Select a Size

Change View
50 EA
R$317.00

About This Item

UNSPSC Code:
41122410
NACRES:
NB.22

R$317.00


Check Cart for Availability

material

polyester film

sterility

non-sterile

packaging

pack of 50 ea (5 x individual-color packs of 10)

manufacturer/tradename

Excel Scientific SP-IDA-10

technique(s)

ELISA: suitable

L

146 mm

W × L

7.94 cm × 14.6 cm

W

79.4 mm

color

assorted colors

Looking for similar products? Visit Product Comparison Guide

General description

SealPlate ColorTab adhesive films consist of 50 μm thickness clear polyester. They minimize evaporation, prevent spillage and contamination between wells, and provide a secure seal for ELISA, EIA, and similar assays. These unique films include a color dot and white writing area on each end tab for plate ID labeling. End tabs with backing removed adhere to the ends of the plate and are perforated so they can be left in place to maintain plate identity even if the center portion of the film is removed. Alternatively, one inscribed color-coded end tab can be removed and pasted in a laboratory notebook while the other remains on the plate for identification. Dimensions 79.4 by 146.0 mm for 96 well plates. Films are supplied with five dot colors (green, lavender, orange, red, blue) in non-sterile single-color packs of 100 and in assorted packs of 50 containing five individual-color packs of 10.

Legal Information

ColorTab is a trademark of Excel Scientific, Inc.
SealPlate is a registered trademark of Excel Scientific, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service