Skip to Content
Merck
All Photos(1)

Key Documents

Z717320

Nichipet EX-Plus II pipette

volume 20-200 μL

Synonym(s):

Nichipet pipette, Precision pipette

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$4,503.00

R$4,503.00


Check Cart for Availability


Select a Size

Change View
1 EA
R$4,503.00

About This Item

UNSPSC Code:
41121500
NACRES:
NB.01

R$4,503.00


Check Cart for Availability

feature

electronic: no
single-channel

packaging

pack of 1 ea
pkg of 1 ea

manufacturer/tradename

Nichiryo 00-NPL02-200

availability

not available in (Not available in France)

volume

20-200 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

The Nichipet EX-Plus ll is a new digital micropipette addition to the Nichiryo family of pipettors. The Nichipet EX-Plus ll offers a new, key feature of one-touch calibration.

Time and money spent on the calibration of pipettes is now greatly reduced!

  • UV resistant
  • Solvent resistant
  • Fully autoclavable (121°C for 20 minutes)
  • One-touch calibration
  • Ceramic pistons
  • Filtered nozzles
  • Easy, convenient locking lever
  • Ergonomic design
  • Single channel, manual, continuously adjustable volume
  • Advanced durability against organic solvent dispensing
  • PTFE (Fluorine resin) in the air tight chamber of the instrument
  • A Perfluoro rubber O-ring with low cubical expansion in the airtight parts
  • Plunger made from a material resistant to organic solvents
  • An alloy spring to avoid metal corrosion by organic solvents
  • Quick and easy to use

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service