Key Documents
Z711012
Media bottles, wide mouth, Lab 45, accessory
polypropylene pour ring
Synonym(s):
240756
Select a Size
12 EA
R$128.00
R$128.00
Estimated to ship onApril 17, 2025
Select a Size
Change View
12 EA
R$128.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.17
R$128.00
Estimated to ship onApril 17, 2025
material
polypropylene pour ring
manufacturer/tradename
WHEATON®
Features and Benefits
• For high temperature applications, use the PBT (polybutylene terephthalate) screw caps and the ETFE (ethylene tetrafluoroethylene) pour lips which can withstand temperatures up to 200ºC up to 2 hours• Autoclavable
Legal Information
WHEATON is a registered trademark of Wheaton Industries, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)