Skip to Content
Merck
All Photos(1)

Key Documents

Z710830

Cap for Tunair shake flask

For 300 mL flask

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
R$163.00

R$163.00


Available to ship onApril 16, 2025Details



Select a Size

Change View
1 EA
R$163.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.22

R$163.00


Available to ship onApril 16, 2025Details


material

polypropylene

description

No lining

sterility

non-sterile

feature

closure type CAP-FLO closure

packaging

case of 1

manufacturer/tradename

IBI SS-3014

Looking for similar products? Visit Product Comparison Guide

General description

Cap for Tunair shake flask is a two-piece cap assembly made up of polypropylene. The filter lining in the cap assembly is replaced by simply pinching the flanges of the inner-closure shell until they snap loose. Then pull it apart and remove the used lining. The lining is then replaced by sandwiching it between two parts of the cap and snapping the cap back together. While cap reassembling, ensure the flanges from the inner piece snap into the mated grooves in the outer piece. This makes the cap assembly stay intact during use. Caps can be cleaned using water with a light detergent solution to remove dirt and contaminants and then air-dry.

Features and Benefits

  • Consists of a two-piece cap for one 300 ml flask.
  • Filter linings are sold separately.
  • Provided in four different colours: red, blue, green, and yellow.
  • Solvent resistant
  • Fully autoclavable flasks and caps.
  • Offers replacable or autoclavable filter linings.

Legal Information

Tunair is a trademark of IBI Scientific

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service