Key Documents
Z709069
ART® non-filtered tip
HLT 300, size 1-300 μL, racked, non-sterile
Synonym(s):
pipet tips, pipette tips
Select a Size
1 PKG
R$1,074.00
R$1,074.00
Estimated to ship onApril 16, 2025
Select a Size
Change View
1 PKG
R$1,074.00
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
maximum volume:
300 μL
size:
1-300 μL
sterility:
non-sterile
feature:
DNase free
RNase free
racked
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 3572
R$1,074.00
Estimated to ship onApril 16, 2025
sterility
non-sterile
feature
DNase free
RNase free
racked
packaging
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename
Molecular BioProducts 3572
maximum volume
300 μL
size
1-300 μL
General description
Ideal for Finnpipette and molded with reference marks at 10, 50 and 100 μl.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.
Legal Information
ART is a registered trademark of Molecular BioProducts, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)