Skip to Content
Merck
All Photos(1)

Key Documents

Z708941

ART® non-filtered tip

HLT 10 Micropoint, maximum volume 10 μL, racked, non-sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing

Select a Size

1 PKG
R$1,159.00
5 PKG
R$4,284.00

R$1,159.00


Check Cart for Availability


Select a Size

Change View
1 PKG
R$1,159.00
5 PKG
R$4,284.00

About This Item

UNSPSC Code:
41121600
NACRES:
NB.01
volume:
10 μL
sterility:
non-sterile
feature:
racked
packaging:
pack of 960 ea (10 racks of 96 tips)
manufacturer/tradename:
Molecular BioProducts 3502

R$1,159.00


Check Cart for Availability

material

colorless

sterility

non-sterile

feature

racked

packaging

pack of 960 ea (10 racks of 96 tips)

manufacturer/tradename

Molecular BioProducts 3502

technique(s)

PCR: suitable

maximum volume

10 μL

volume

10 μL

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Micropoint design and 2 μl reference mark make this tip ideal for low volume pipetting.
Molecular BioProducts Pure lines of pipette tips are pre-sterilized and packaged in a controlled environment to eliminate contamination. These tips are tested by an independent lab to ensure that they meet industry standards. It is ideal for a wide variety of applications.

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service