Skip to Content
Merck
All Photos(1)

Documents

Z706345

Millipore

Amicon Ultra centrifugal filter units

Ultra-15, MWCO 10 kDa

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900

material

polyethylene cap
polypropylene tube
regenerated cellulose membrane
styrene-butadiene housing

sterility

non-sterile

manufacturer/tradename

Millipore UFC901008

concentrate volume

200 μL

membrane area × diam. × L

7.6 cm2 × 35 mm × 121 mm

tube size

35 x 121 mm

working volume

~15 mL

pore size

10 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

Typical protein recovery 93.03%
High recovery centrifugal filters for concentrating samples. Can be used to concentrate biological samples containing antigens, antibodies, enzymes, nucleic acids or micoorganisms. Excellen for use to purify macromolecular components from cell lysates. Ideal for desalting and buffer exchanges.
Swing bucket rotorL 4,000 x g; Fixed angle rotor (35°): 5,000 x g.
  • High recovery Ultracel regenerated cellulose membrane in a range of molecular weight cut-offs.
  • High retentate recovery of >90%
  • Vertical membrane reduces concentration polarization for ultra-fast spin times (as fast as 10-15 mins)
  • Heat-sealed membrane minimizes downstream extractables
  • 100% integrity tested for reliable performance
  • Convenient sample monitoring with translucent housing and volume graduations
  • Direct pipettor sample access eliminates processing step to recover concentrate
  • High concentration factors of 80 - 100X

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service