Key Documents
Z659738
Bandelin SONOREX Ultrasonic bath accessories
insert tub with lid for Z659673 and Z659029
Select a Size
1 EA
R$510.00
R$510.00
Estimated to ship onJune 01, 2025
Select a Size
Change View
1 EA
R$510.00
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
R$510.00
Estimated to ship onJune 01, 2025
material
polyethylene
packaging
pack of 1 ea
manufacturer/tradename
Bandelin KW 5
availability
not available in USA and Canada
L × W × D
254 mm × 96 mm × 130 mm
General description
For use with chemically aggressive liquids, removing chemically aggressive or abrasive contamination or sonicating sample liquids, insert cups are used (indirect sonication). Because nothing may be placed on the bottom of the ultrasonic bath.
Features and Benefits
  • for chemically aggressive liquids
  • protects the stainless steel tub
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)