Z364886
Wheaton media bottles, glass
capacity 500 mL, PTFE lined cap
Synonym(s):
Wheaton bottles, laboratory bottles, media bottle
Select a Size
R$3,690.00
Select a Size
About This Item
R$3,690.00
Recommended Products
material
PTFE cap
borosilicate glass bottle
round bottle
cap size
33-430
Joint
Joints threaded neck (33-430)
description
Autoclavable with cap loose.
sterility
non-sterile
feature
graduations
with cap
packaging
pack of 24 ea
manufacturer/tradename
WHEATON® 219819
H × diam.
188 mm × 88 mm , Cap off size
capacity
500 mL
Looking for similar products? Visit Product Comparison Guide
General description
- Wheaton media bottles are manufactured from low-alkali borosilicate glass.
- Clear bottles are ideal for storing culture media.
- No-drip lip for clean pouring.
- Sloping shoulder for easier cleaning.
- Marking patch for identification.
- Graduations show approximate volume.
- Includes an autoclavable cap for sample protection.
Features and Benefits
- With graduations and marking patch for easy measuring volume and identifying content
- Made from low extractable, clear borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
- Hand grips on 1 Liter bottle provide easy handling
- Special pour lip facilitates drip free pouring
- Packaged in partitioned carton to help prevent scratch marks on bottles
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service