Key Documents
Z362816
Nalgene® Polymethylpentene Erlenmeyer flask
capacity 500 mL
Synonym(s):
conical flasks, erlenmeyer flasks, flask erlenmeyer, flasks erlenmeyer
Select a Size
4 EA
R$1,739.00
R$1,739.00
Check Cart for Availability
Select a Size
Change View
4 EA
R$1,739.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
R$1,739.00
Check Cart for Availability
material
colorless polypropylene cap
flat bottom
polymethylpentene
wide-neck
Joint
Joints neck (43)
sterility
non-sterile
feature
closure type Screw cap
with cap
packaging
pack of 4
manufacturer/tradename
Nalgene 4109-0500
capacity
500 mL
General description
Excellent combination of autoclavability, chemical resistance, impact strength, and clarity. Transparent to microwave radiation. Polypropylene screw closure.
Other Notes
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)