Z359947
Pellet pestles
blue polypropylene (Autoclavable)
Synonym(s):
749521-1500
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121800
eCl@ss:
32080115
NACRES:
NB.22
Recommended Products
material
blue polypropylene (Autoclavable)
manufacturer/tradename
Kimble®
Looking for similar products? Visit Product Comparison Guide
General description
Hand-operated or motor-driven grinders for resuspending pellets or disrupting soft tissue in microcentrifuge tubes. Pestle ends are specially designed to mate with 0.5 mL or 1.5 mL microtubes. Shaft is convenient length and diameter for gentle manual back-and-forth rotation or more vigorous motorized operation.
Z359955 will fit the cordless motor product Z359971 only by using the replacement adapter Z359998 for the motor, which is purchased separately.
Compatibility
Fits 1.5 mL microtube
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Gesine Kaiser et al.
Malaria journal, 16(1), 305-305 (2017-08-03)
Malaria research is greatly dependent on and has drastically advanced with the possibility of genetically modifying Plasmodium parasites. The commonly used transfection protocol by Janse and colleagues utilizes blood stage-derived Plasmodium berghei schizonts that have been purified from a blood
Nonno Hasegawa et al.
BMC genomics, 22(1), 54-54 (2021-01-16)
The honey bee parasite, Varroa destructor, is a leading cause of honey bee population declines. In addition to being an obligate ectoparasitic mite, Varroa carries several viruses that infect honey bees and act as the proximal cause of colony collapses.
T Peter Kingham et al.
Oncotarget, 8(12), 18821-18831 (2016-12-10)
Resection of colorectal liver metastasis (CRLM) can be curative. Predicting which patients may benefit from resection, however, remains challenging. Some microRNAs (miRNAs) become deregulated in cancers and contribute to cancer progression. We hypothesized that miRNA expression can serve as a
Moshe Gross et al.
Stress (Amsterdam, Netherlands), 1-10 (2018-04-13)
Prenatal stress may increase concentrations of maternal glucocorticoids, which restrict fetal growth, with variable impact upon postnatal development. Among key regulators of stress hormone effects are the glucocorticoid receptor (GR) and 11β-hydroxysteroid dehydrogenase-2 (11βHSD2), the enzyme that inactivates glucocorticoid. This
Aniek van der Vaart et al.
Science advances, 6(21), eaay3823-eaay3823 (2020-06-05)
SWI/SNF (switch/sucrose nonfermenting) complexes regulate transcription through chromatin remodeling and opposing gene silencing by Polycomb group (PcG) proteins. Genes encoding SWI/SNF components are critical for normal development and frequently mutated in human cancer. We characterized the in vivo contributions of
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service