Key Documents
Z332313
Bottle adapters for BRAND® Dispensers and Burettes
GL 45 thread, S 60 bottle, PE
Select a Size
1 EA
R$335.00
R$335.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$335.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.01
R$335.00
Check Cart for Availability
material
PE
manufacturer/tradename
BRAND 704348
fitting
S 60 bottle
GL 45 thread
Related Categories
General description
For all Dispensette® models as well as seripettor® and QuikSip
Other Notes
BR704348 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Dispensette is a registered trademark of BRAND GMBH + CO KG
QuikSip is a trademark of BRAND GMBH + CO KG
seripettor is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)