Z325309
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 2 mL, two marks
All Photos(1)
About This Item
Recommended Products
material
AR-Glas®
description
partial delivery
feature
class AS
packaging
pack of 12 ea
manufacturer/tradename
BRAND 29723
volume measuring range
2 mL, accuracy: 0.010 mL
capacity
2 mL
Related Categories
General description
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
According to DIN EN ISO 648.
Other Notes
BR29723 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.