Skip to Content
Merck
All Photos(2)

Key Documents

WH0388591M2

Sigma-Aldrich

Monoclonal Anti-FLJ46380 antibody produced in mouse

clone 1C5, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FLJ46380 protein

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,068.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Check Cart for Availability

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1C5, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunofluorescence: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RNF207(388591)

General description

FLJ46380 is also known as ring finger protein 207 (RNF207). It is an E3 ligase expressed in the heart. The gene encoding it is located on human chromosome 1p36.31.

Immunogen

FLJ46380 (NP_997279, 135 a.a. ~ 244 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
HVFRGRPGSGFSSTSLGHLGPKCEPHYTGGETEVQNKGLEPVSRQWQRLRPFDLGRAHWSPIQGGVVDLHRRGSPVCRPGPTLKGLCYPSGIEAATAQGRWGQHAVPSGL

Biochem/physiol Actions

FLJ46380 or ring finger protein 207 (RNF207) has been studied as a marker for acute myocardial infarction.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiu-Yue Han et al.
Clinical science (London, England : 1979), 128(11), 751-760 (2015-01-20)
Ubiquitin ligase (E3) is a decisive element of the ubiquitin-proteasome system (UPS), which is the main pathway for intracellular protein turnover. Recently, circulating E3 ligases have been increasingly considered as cancer biomarkers. In the present study, we aimed to determine
Christopher Newton-Cheh et al.
Nature genetics, 41(4), 399-406 (2009-03-24)
QT interval duration, reflecting myocardial repolarization on the electrocardiogram, is a heritable risk factor for sudden cardiac death and drug-induced arrhythmias. We conducted a meta-analysis of three genome-wide association studies in 13,685 individuals of European ancestry from the Framingham Heart

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service