Skip to Content
Merck
All Photos(5)

Key Documents

WH0170691M1

Sigma-Aldrich

Monoclonal Anti-ADAMTS17 antibody produced in mouse

clone 3B7, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-ADAM metallopeptidase with thrombospondin type 1 motif, 17, Anti-FLJ32769

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,068.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3B7, monoclonal

form

buffered aqueous solution

species reactivity

human, mouse, rat

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

This gene encodes a member of the ADAMTS (a disintegrin and metalloproteinase with thrombospondin motifs) protein family. ADAMTS family members share several distinct protein modules, including a propeptide region, a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 (TS) motif. Individual members of this family differ in the number of C-terminal TS motifs, and some have unique C-terminal domains. The protein encoded by this gene has a high sequence similarity to the protein encoded by ADAMTS19, another family member. The function of this protein has not been determined. (provided by RefSeq)

Immunogen

ADAMTS17 (NP_620688, 543 a.a. ~ 650 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DGDWSPWGAWSMCSRTCGTGARFRQRKCDNPPPGPGGTHCPGASVEHAVCENLPCPKGLPSFRDQQCQAHDRLSPKKKGLLTAVVVDDKPCELYCSPLGKESPLLVAD

Biochem/physiol Actions

ADAM metallopeptidase with thrombospondin type 1 motif 17 (ADAMTS17) is associated with Weill-Marchesani-like syndrome. Knockdown of the protein has been shown to prevent the growth of breast cancer cells in vitro. ADAMTS17 binds to the extracellular matrix.[1]

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mohd Hussain Shah et al.
Molecular vision, 20, 790-796 (2014-06-19)
Weill-Marchesani syndrome (WMS) is a rare connective tissue disorder, characterized by short stature, microspherophakic lens, and stubby hands and feet (brachydactyly). WMS is caused by mutations in the FBN1, ADAMTS10, and LTBP2 genes. Mutations in the LTBP2 and ADAMTS17 genes
Tao Zhou et al.
Experimental and therapeutic medicine, 14(2), 1131-1135 (2017-08-16)
The aim of the study was to investigate the mechanism of microRNA (miR)-146a-mediated activation of interleukin-6/signal transducer and activator of transcription 3 (IL-6/STAT3) in lumbar intervertebral disc degeneration. To obtain intervertebral tissue, we recruited 5 patients with lumbar intervertebral disc
Zanhui Jia et al.
Journal of cellular biochemistry, 115(10), 1829-1839 (2014-06-07)
Adamts17 is a member of a family of secreted metalloproteinases. In this report, we show that knockdown of Adamts17 expression induces apoptosis and inhibits breast cancer cell growth. Adamts17 expression can rapidly be induced by estrogens. siRNA knockdown of Sp1

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service