Skip to Content
Merck
All Photos(4)

Key Documents

WH0064787M1

Sigma-Aldrich

Monoclonal Anti-EPS8L2 antibody produced in mouse

clone 6C2, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-EPS8-like 2, Anti-EPS8R2, Anti-FLJ21935, Anti-FLJ22171, Anti-MGC126530, Anti-MGC3088

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Check Cart for Availability


Select a Size

Change View
100 μG
R$3,068.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Check Cart for Availability

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6C2, monoclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EPS8L2(64787)

General description

This gene encodes a member of the EPS8 gene family. The encoded protein, like other members of the family, is thought to link growth factor stimulation to actin organization, generating functional redundancy in the pathways that regulate actin cytoskeletal remodeling. (provided by RefSeq)

Immunogen

EPS8L2 (NP_073609, 615 a.a. ~ 714 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
ERSQPVSQPLTYESGPDEVRAWLEAKAFSPRIVENLGILTGPQLFSLNKEELKKVCGEEGVRVYSQLTMQKAFLEKQQSGSELEELMNKFHSMNQRRGED

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service