Skip to Content
Merck
All Photos(2)

Key Documents

WH0057717M3

Sigma-Aldrich

Monoclonal Anti-PCDHB16 antibody produced in mouse

clone 3H1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-KIAA1621, Anti-ME1, Anti-PCDH3X, Anti-PCDHB8a, Anti-PCDHBETA16, Anti-protocadherin beta 16

Sign Into View Organizational & Contract Pricing

Select a Size

50 μG
R$3,068.00

R$3,068.00


Estimated to ship onMay 04, 2025



Select a Size

Change View
50 μG
R$3,068.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Estimated to ship onMay 04, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3H1, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PCDHB16(57717)

General description

This gene is a member of the protocadherin beta gene cluster, one of three related gene clusters tandemly linked on chromosome five. The gene clusters demonstrate an unusual genomic organization similar to that of B-cell and T-cell receptor gene clusters. The beta cluster contains 16 genes and 3 pseudogenes, each encoding 6 extracellular cadherin domains and a cytoplasmic tail that deviates from others in the cadherin superfamily. The extracellular domains interact in a homophilic manner to specify differential cell-cell connections. Unlike the alpha and gamma clusters, the transcripts from these genes are made up of only one large exon, not sharing common 3′ exons as expected. These neural cadherin-like cell adhesion proteins are integral plasma membrane proteins. Their specific functions are unknown but they most likely play a critical role in the establishment and function of specific cell-cell neural connections. (provided by RefSeq)

Immunogen

PCDHB16 (AAH36062.1, 1 a.a. ~ 776 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEIGWMHNRRQRQVLVFFVLLSLSGAGAELGSYSVVEETERGSFVANLGKDLGLGLTEMSTRKARIISQGNKQHLQLKAQTGDLLINEKLDREELCGPTEPCILHFQVLMENPLEIFQAELRVIDINDHSPMFTEKEMILKIPENSPLGTEFPLNHALDLDVGSNNVQNYKISPSSHFRVLIHEFRDGRKYPELVLDKELDREEEPQLRLTLTALDGGSPPRSGTAQVRIEVVDINDNAPEFEQPIYKVQIPENSPLGSLVATVSARDLDGGANGKISYTLFQPSEDISKTLEVNPMTGEVRLRKQVDFEMVTSYEVRIKATDGGGLSGKCTLLLQVVDVNDNPPQVTMSALTSPIAENSPEIVVAVFSVSDPDSGNNGKTISSIQEDLPFLLKPSVKNFYTLVTERALDREARAEYNITLTVTDMGTPRLKTEHNITVQISDVNDNAPTFTQTSYTLFVRENNSPALHIGSVSATDRDSGTNAQVTYSLLPPQDPHLPLASLVSINADNGHLFALRSLDYEALQAFEFRVGATDRGSPALSREALVRVLVLDANDNSPLVLYPLQNGSAPCTELVPRAAEPGYLVTKVVAVDGDSGQNAWLSYQLLKATEPGLFGVWAHNGEVRTARLLSERDAAKQRLVVLVKDNGEPPRSATATLHVLLVDGFSQPFLPLPEAAPGQTQANSLTVYLVVALASVSSLFLFSVLLFVAVRLCRRSRAASVGRCSMPEGPFPGRLVDVSGTGTLSQSYQYEVCLTGGSETSEFKFLKPIIPNFSP

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service