Skip to Content
Merck
All Photos(6)

Key Documents

WH0005307M1

Sigma-Aldrich

Monoclonal Anti-PITX1 antibody produced in mouse

clone 5G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-BFT, Anti-POTX, Anti-PTX1, Anti-paired-like homeodomain transcription factor 1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Estimated to ship onApril 23, 2025



Select a Size

Change View
100 μG
R$3,068.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Estimated to ship onApril 23, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5G4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PITX1(5307)

Related Categories

General description

This gene encodes a member of the RIEG/PITX homeobox family, which is in the bicoid class of homeodomain proteins. Members of this family are involved in organ development and left-right asymmetry. This protein acts as a transcriptional regulator involved in basal and hormone-regulated activity of prolactin. (provided by RefSeq)

Immunogen

PITX1 (NP_002644, 225 a.a. ~ 313 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPSSMGPGAVPGMPNSGLNNINNLTGSSLNSAMSPGACPYGTPASPYSVYRDTCNSSLASLRLKSKQHSSFGYGGLQGPASGLNACQYN

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service