Skip to Content
Merck
All Photos(3)

Documents

WH0003576M5

Sigma-Aldrich

Monoclonal Anti-IL8 antibody produced in mouse

clone 6G4, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-CXCL8, Anti-GCP1, Anti-Interleukin 8, Anti-K60, Anti-NAP1, Anti-SCYB8, Anti-TSG1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

6G4, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IL8(3576)

Related Categories

General description

Interleukin-8 (IL-8)/CXCL8 is an important chemoattractant cytokine and activator of neutrophils. It is liberated from endothelial cells, gingival fibroblasts, neutrophils, monocytes and phagocytes in the gingival crevice. IL-8 belongs to the CXC subfamily. This gene is located on human chromosome 4q13.3.
The protein encoded by this gene is a member of the CXC chemokine family. This chemokine is one of the major mediators of the inflammatory response. This chemokine is secreted by several cell types. It functions as a chemoattractant, and is also a potent angiogenic factor. This gene is believed to play a role in the pathogenesis of bronchiolitis, a common respiratory tract disease caused by viral infection. This gene and other ten members of the CXC chemokine gene family form a chemokine gene cluster in a region mapped to chromosome 4q. (provided by RefSeq)

Immunogen

IL8 (AAH13615.1, 21 a.a. ~ 99 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
EGAVLPRSAKELRCQCIKTYSKPFHPKFIKELRVIESGPHCANTEIIVKLSDGRELCLDPKENWVQRVVEKFLKRAENS

Biochem/physiol Actions

Interleukin-8 (IL-8) chemoattracts and activates neutrophils. It is involved in guiding neutrophils and oligodendrocytes to sites of infection. The protein also stimulates angiogenesis and tumor growth. It is also associated with inflammatory diseases.
Interleukin-8 (IL-8)/CXCL8 participates in the progression of peripheral muscle weakness in individuals with COPD (chronic obstructive pulmonary disease). It helps in the transport of neutrophils across endothelium, pulmonary epithelium and fibroblasts. CXCL8 induces adhesion of neutrophils to extracellular matrix proteins and cytokine-stimulated endothelial monolayers via interaction with CD11b/CD18 (β2-integrins).

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pathophysiological roles of interleukin-8/CXCL8 in pulmonary diseases
Mukaida N.
American Journal of Physiology. Lung Cellular and Molecular Physiology, 284(4), L566-L577 (2003)
Satsuki Shimizu et al.
Scientific reports, 13(1), 5041-5041 (2023-03-29)
Infantile skin problems not only cause temporary pain and discomfort, but also have a long-term impact on health. Hence, the purpose of this cross-sectional study was to clarify the relationship between inflammatory cytokines and Malassezia fungal facial skin problems in
Muscle force during an acute exacerbation in hospitalised patients with COPD and its relationship with CXCL8 and IGF-I
Spruit MA, et al.
Thorax, 58(9), 752-756 (2003)
Association between interleukin-8 levels and chronic periodontal disease: A PRISMA-compliant systematic review and meta-analysis
Finoti LS, et al.
Medicine, 96(22) (2017)
The chemokine and chemokine receptor superfamilies and their molecular evolution.
Zlotnik A
Genome Biology, 7(12), 243-243 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service