Skip to Content
Merck
All Photos(3)

Key Documents

WH0002395M3

Sigma-Aldrich

Monoclonal Anti-FXN antibody produced in mouse

clone 3G9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-FA, Anti-FARR, Anti-FRDA, Anti-MGC57199, Anti-X25, Anti-frataxin

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$4,997.00

R$4,997.00


Estimated to ship onApril 29, 2025



Select a Size

Change View
100 μG
R$4,997.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$4,997.00


Estimated to ship onApril 29, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

3G9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG1κ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FXN(2395)

General description

Frataxin (FXN) protein consists of α/β fold, which is followed by the C-terminal region (CTR), with a nonperiodic structure.The gene is located on human chromosome 9q21.11.

Immunogen

FXN (AAH48097.1, 91 a.a. ~ 200 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
DETTYERLAEETLDSLAEFFEDLADKPYTFEDYDVSFGSGVLTVKLGGDLGTYVINKQTPNKQIWLSSPSSGPKRYDWTGKNWVYSHDGVSLHELLAAELTKALKTKLDL

Biochem/physiol Actions

Frataxin (FXN) is involved in the activation of iron-sulfur cluster assembly.Overexpression of FXN in reticulocytes is associated with oxidative stress and iron status.Deficiency of FXN in human astrocytes is linked to cell death and release of neurotoxins.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Frataxin expression in reticulocytes of non-splenectomized and splenectomized patients with HbE-beta-thalassaemia
Suebpeng, et al.
Clinical Biochemistry, 49(6), 463-466 (2016)
Evidence for chromosome fragility at the frataxin locus in Friedreich ataxia
Kumari,, et al.
Mutation Research. Fundamental and Molecular Mechanisms of Mutagenesis, 781, 14-21 (2015)
The alteration of the C-terminal region of human frataxin distorts its structural dynamics and function
Faraj, S, et al.
FEBS Journal, 281(15), 3397-3419 (2014)
Insights on the conformational dynamics of human frataxin through modifications of loop-1.
Noguera ME, et al.
Archives of Biochemistry and Biophysics, 636, 123-137 (2017)
Frida Loría et al.
Neurobiology of disease, 76, 1-12 (2015-01-03)
Friedreich's ataxia (FA) is a recessive, predominantly neurodegenerative disorder caused in most cases by mutations in the first intron of the frataxin (FXN) gene. This mutation drives the expansion of a homozygous GAA repeat that results in decreased levels of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service