Skip to Content
Merck
All Photos(4)

Key Documents

WH0001787M1

Sigma-Aldrich

Monoclonal Anti-DNMT2 antibody produced in mouse

clone 1E12, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-DNA (cytosine-5-)-methyltransferase 2, Anti-M.HsaIIP, Anti-PuMet

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$3,068.00

R$3,068.00


Estimated to ship onApril 23, 2025



Select a Size

Change View
100 μG
R$3,068.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

R$3,068.00


Estimated to ship onApril 23, 2025


biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1E12, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
indirect immunofluorescence: suitable
western blot: 1-5 μg/mL

isotype

IgG1λ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TRDMT1(1787)

General description

This gene encodes a protein responsible for the methylation of aspartic acid transfer RNA. While similar in sequence and structure to DNA cytosine methyltransferases, this gene is distinct and highly conserved in its function among taxa. Alternative splicing results in multiple transcript variants encoding different isoforms. (provided by RefSeq)

Immunogen

DNMT2 (AAH47733, 1 a.a. ~ 391 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MEPLRVLELYSGVGGMHHALRESCIPAQVVAAIDVNTVANEVYKYNFHHTQLLAKTIEGITLEEFDRLSFDMILMSPPCQPFTRIGRQGDMTDSRTNSFLYILDILPRLQKLPKYILLENVKGFEVSSTRDLLIQTIENCGFQYQEFLLSPTSLGIPNSRLRYFLIAKLQSEPLPFQAPGQVLMEFPKIESVHPQKYAMDVEEKIQEKNVEPNISFDGSIQCSGKDAILFKLETAEEIHRKNQQDSDLSVKMLKDFLEDDTDVNQYLLPPKSLLRYALLLDIVQPTCRRSVCFTKGYGSYIEGTGSVLQTAEDVQVENIYKSLTNLSQEEQITKLLILKLRYFTPKEIANLLGFPPEFGFPEKITVKQRYRLLGNSLNVHVVAKLIKILYE

Biochem/physiol Actions

DNA methyltransferase 2 (DNMT2) might be involved in de novo methylation of newly integrated retroviral DNA in mammalian DNA. DNMT2 functions as a tRNA methyltransferase in lower eukaryotes. DNMT2 plays an essential role in zebrafish development by enhancing cytoplasmic RNA methylation. DNMT2 promotes DNA methylation in Drosophila.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ayala Tovy et al.
PLoS pathogens, 6(2), e1000775-e1000775 (2010-02-23)
Cytosine-5 methyltransferases of the Dnmt2 family function as DNA and tRNA methyltransferases. Insight into the role and biological significance of Dnmt2 is greatly hampered by a lack of knowledge about its protein interactions. In this report, we address the subject
J A Yoder et al.
Human molecular genetics, 7(2), 279-284 (1998-03-21)
Trace levels of 5-methylcytosine persist in the DNA of mouse embryonic stem cells that are homozygous for null mutations in Dnmt1 , the gene for the one previously recognized metazoan DNA methyltransferase. This residual 5-methylcytosine may be the product of
Natascha Kunert et al.
Development (Cambridge, England), 130(21), 5083-5090 (2003-08-29)
The methylation status of Drosophila DNA has been discussed controversially over a long time. Recent evidence has provided strong support for the existence of 5-methylcytosine in DNA preparations from embryonic stages of fly development. The Drosophila genome contains a single

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service